Learn More
TARGETMOL CHEMICALS INC PDK4-IN-1 hydrochloride 1MG
SDP
Catalog No. 502875923
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. PDK4-IN-1 hydrochloride, an anthraquinone derivative, is a potent and orally active inhibitor of pyruvate dehydrogenase kinase 4 (PDK4) with an IC50 value of 84 nM. Purity 99.54%
Catalog No. 50-287-5923 Supplier TARGETMOL CHEMICALS INC Supplier No. T12412L1MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. PDK4-IN-1 hydrochloride, an anthraquinone derivative, is a potent and orally active inhibitor of pyruvate dehydrogenase kinase 4 (PDK4) with an IC50 value of 84 nM. Purity 99.54%