Learn More
Medchemexpress LLC HY-135954A 5mg Medchemexpress, PDK4-IN-1 (hydrochloride) CAS:2310262-11-2 Purity:>98%
SDP
Catalog No. 502030387
Encompass_Preferred
Medchemexpress, HY-135954A 5mg PDK4-IN-1 (hydrochloride) CAS:2310262-11-2 PDK4-IN-1 hydrochloride is an anthraquinone derivative and a potent and orally active pyruvate dehydrogenase kinase 4 (PDK4) inhibitor with an IC50 value of 84 nM. PDK4-IN-1 hydrochloride potently represses cellular transformation and cellular proliferation and induces apoptosis. PDK4-IN-1 hydrochloride has antidiabetic, anticancer and anti-allergic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0387 Supplier Medchemexpress LLC Supplier No. HY135954A5MG
Medchemexpress, HY-135954A 5mg PDK4-IN-1 (hydrochloride) CAS:2310262-11-2 PDK4-IN-1 hydrochloride is an anthraquinone derivative and a potent and orally active pyruvate dehydrogenase kinase 4 (PDK4) inhibitor with an IC50 value of 84 nM. PDK4-IN-1 hydrochloride potently represses cellular transformation and cellular proliferation and induces apoptosis. PDK4-IN-1 hydrochloride has antidiabetic, anticancer and anti-allergic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.