Learn More
Enzo Life Sciences PDKtide (biotinylated) (100µg)
SDP
Supplier:  Enzo Life Sciences BMLP2510100
Encompass_Preferred
This peptide, Biotin-Ahx-KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC, is derived from AKT1 and PKN2/PRK2. It is a substrate for PDK1, CDK9/Cyclin T1 and JAK2.The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes. Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1315