Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDL1 Detection Set (Risk Free), ProSci
SDP

Catalog No. 16595556 Shop All ProSci Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-556 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-556 Supplier ProSci LLC Supplier No. RF16030SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Detection Sets

PDL1 Monoclonal specifically detects PDL1 in Human samples. It is validated for ELISA, Western Blot, ICC, IHC-P, Flow, IF

Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antigen-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PD-L1 and PD-L2. PD-L1 is a B7-related protein that inhibits cell-mediated immune responses by reducing the secretion of IL-2 and IL-10 from memory T cells. This suggests that PD-L1 may be useful in reducing allogenic CD4+ memory T-cell responses to endothelial cells, thereby reducing the likelihood of host immune responses to allografts. ProSci's Risk-FreeTM antibodies are mouse monoclonal antibodies made to improve in vivo studies. Unlike antibodies developed using proteins made in yeast or bacteria, Risk-FreeTM antibodies are developed with antigens expressed in mammalian cell lines, giving the most native post-translational modifications to the protein. Validated for flow cytometry and ELISA Rigorously tested for the following applications: Immunoblot Immunohistochemistry Immunocytochemistry Immunofluorescence Multiple antibodies per target allowing the user to choose the best antibody for their application Available individually or as a set Risk-FreeTM means they are guaranteed to work for their approved applications

Specifications

Antigen PDL1
Applications ELISA, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence
Classification Monoclonal
Concentration Antibody 1 mg/mL
Conjugate Unconjugated
Formulation PBS containing 0.02% sodium azide.
Host Species Mouse
Purification Method Antibodies are supplied as protein A purified IgG1.
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Antibodies can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Detection Sets
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.