Learn More
Cayman Chemical PDL1 EXrAClular DomaInh 100ug
SDP
Catalog No. 502941270
Encompass_Preferred
Source: Recombinant C-terminal mouse IgG1 Fc-tagged human PD-L1 expressed in HEK293 cells • Amino acids: 19-239 • MW: 51.7 kDa
Catalog No. 50-294-1270 Supplier Cayman Chemical Supplier No. 28378100 UG
Source: Recombinant C-terminal mouse IgG1 Fc-tagged human PD-L1 expressed in HEK293 cells • Amino acids: 19-239 • MW: 51.7 kDa