Learn More
Sino Biological PD-L1/B7-H1/CD274 Antibody (PE), Rabbit MAb 25Tests
Supplier:  Sino Biological 10084R312P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PD-L1/B7-H1/CD274 (rh PD-L1/B7-H1/CD274; Catalog#10084-H08H; NP_054862.1; Met1-Thr239) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-224-7565