Learn More
Cayman Chemical PDL1CD274 RabbIt MoncLNal 25
SDP
Catalog No. 502915261
Encompass_Preferred
Immunogen: Recombinant mouse PD-L1 • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) PD-L1 • Applications: FC
Catalog No. 50-291-5261 Supplier Cayman Chemical Supplier No. 3706525 TEST
Immunogen: Recombinant mouse PD-L1 • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) PD-L1 • Applications: FC