Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque PD-L1 / B7-H1 Protein, Fc Tag;HEK293;1MG;PD1,PDL1,PD-L1,CD274,B7-H1,B7H1,B7-H,B7H,PDCD1L1,PDCD1LG1
Catalog No. 501659102
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque PD-L1 / B7-H1 Protein, Fc Tag;1MG;Cynomolgus / Rhesus macaque PD-L1, Fc Tag (PD1-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # G7PSE7).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.8 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus PD-1, His Tag (Cat. No. PD1-C5223) at 10 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque PD-L1, Fc Tag (Cat. No. PD1-C5253) with a linear range of 0.156-5 μg/mL (QC tested).
Catalog No. 50-165-9102 Supplier ACROBiosystems Supplier No. PD1C52531MG
Recombinant Protein;Cynomolgus / Rhesus macaque PD-L1 / B7-H1 Protein, Fc Tag;1MG;Cynomolgus / Rhesus macaque PD-L1, Fc Tag (PD1-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # G7PSE7).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.8 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus PD-1, His Tag (Cat. No. PD1-C5223) at 10 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque PD-L1, Fc Tag (Cat. No. PD1-C5253) with a linear range of 0.156-5 μg/mL (QC tested).