Learn More
ACROBiosystems Biotinylated Human PD-L1 Protein, Fc-Avitag, Lyophilized, 200 μg
Catalog No. 501658214
Encompass_Preferred
Programmed death-ligand 1 (PD-L1), also known as B7-H1, is a member of the B7 family of immune molecules involved in regulating cellular and humoral immune responses. This biotinylated human PD-L1 protein is expressed in human 293 cells (HEK293) and contains amino acids Phe 19 to Arg 238. It features a human IgG1 Fc tag at the C-terminus, followed by an Avi tag for enzymatic biotinylation. The protein is supplied as a lyophilized powder and is suitable for various immunological applications.
  • Enzymatically biotinylated using a site-specific labeling technology
  • High purity: >90% by SDS-PAGE and >95% by SEC-MALS
  • Calculated molecular weight of 53.6 kDa
  • Validated via ELISA for binding to PD-1 and anti-PD-L1 monoclonal antibodies
  • Lyophilized formulation includes trehalose as a protectant
Catalog No. 50-165-8214 Supplier ACROBiosystems Supplier No. PDLH82F2200UG
Programmed death-ligand 1 (PD-L1), also known as B7-H1, is a member of the B7 family of immune molecules involved in regulating cellular and humoral immune responses. This biotinylated human PD-L1 protein is expressed in human 293 cells (HEK293) and contains amino acids Phe 19 to Arg 238. It features a human IgG1 Fc tag at the C-terminus, followed by an Avi tag for enzymatic biotinylation. The protein is supplied as a lyophilized powder and is suitable for various immunological applications.
  • Enzymatically biotinylated using a site-specific labeling technology
  • High purity: >90% by SDS-PAGE and >95% by SEC-MALS
  • Calculated molecular weight of 53.6 kDa
  • Validated via ELISA for binding to PD-1 and anti-PD-L1 monoclonal antibodies
  • Lyophilized formulation includes trehalose as a protectant