Learn More
ACROBiosystems Recombinant Protein;Human PD-L1 / B7-H1 Protein, Llama IgG2b Fc Tag, low endotoxin;HEK293;100UG;PDL,PDL1,PD-L1,CD274,B7-H1,B7H1,B7-H,B7H,PDCD1L1,PDCD1LG1
Catalog No. 501657944
Encompass_Preferred
Recombinant Protein;Human PD-L1 / B7-H1 Protein, Llama IgG2b Fc Tag, low endotoxin;100UG;Human PD-L1, Llama IgG2b Fc Tag, low endotoxin (PDL-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19- Arg 238 (Accession # Q9NZQ7-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 53.0 kDa. The protein migrates as 65-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-1, His Tag (Cat. No. PD1-H5221) at 1 μg/mL (100 μL/well) can bind Human PD-L1, Llama IgG2b Fc Tag, low endotoxin (Cat. No. PDL-H5250) with a linear range of 0.078-2.5 μg/mL (QC tested).;Immobilized Human PD-L1, Llama IgG2b Fc Tag, low endotoxin (Cat. No. PDL-H5250) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Fc Tag (Cat. No. PD1-H82F1) with a linear range of 20-313 ng/mL (Routinely tested).
Catalog No. 50-165-7944 Supplier ACROBiosystems Supplier No. PDLH5250100UG
Recombinant Protein;Human PD-L1 / B7-H1 Protein, Llama IgG2b Fc Tag, low endotoxin;100UG;Human PD-L1, Llama IgG2b Fc Tag, low endotoxin (PDL-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19- Arg 238 (Accession # Q9NZQ7-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 53.0 kDa. The protein migrates as 65-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-1, His Tag (Cat. No. PD1-H5221) at 1 μg/mL (100 μL/well) can bind Human PD-L1, Llama IgG2b Fc Tag, low endotoxin (Cat. No. PDL-H5250) with a linear range of 0.078-2.5 μg/mL (QC tested).;Immobilized Human PD-L1, Llama IgG2b Fc Tag, low endotoxin (Cat. No. PDL-H5250) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Fc Tag (Cat. No. PD1-H82F1) with a linear range of 20-313 ng/mL (Routinely tested).