Learn More
ACROBiosystems Recombinant Protein;Human PD-L1 / B7-H1 Protein, Strep Tag;HEK293;250UG;PD1,PDL1,PD-L1,CD274,B7-H1,B7H1,B7-H,B7H,PDCD1L1,PDCD1LG1
Catalog No. 501658411
Encompass_Preferred
Recombinant Protein;Human PD-L1 / B7-H1 Protein, Strep Tag;250UG;Human PD-L1, Strep Tag (PD1-H5282) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 35-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1, Strep Tag (Cat. No. PD1-H5282) at 5 μg/mL ( 100 μl/well ) can bind Biotinylated Human PD-1, His Tag & Fc tag (Cat. No. PD1-H82F2) with a linear range of 2-60 ng/mL (QC tested).
Catalog No. 50-165-8411 Supplier ACROBiosystems Supplier No. PD1H5282250UG
Recombinant Protein;Human PD-L1 / B7-H1 Protein, Strep Tag;250UG;Human PD-L1, Strep Tag (PD1-H5282) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 35-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1, Strep Tag (Cat. No. PD1-H5282) at 5 μg/mL ( 100 μl/well ) can bind Biotinylated Human PD-1, His Tag & Fc tag (Cat. No. PD1-H82F2) with a linear range of 2-60 ng/mL (QC tested).