Programmed death-ligand 2 (PD-L2), also known as CD273, is a member of the immunoglobulin superfamily that plays a critical role in regulating T-cell responses. This human PD-L2 protein is expressed in HEK293 cells with a human IgG1 Fc tag at the C-terminus and features low endotoxin levels for reliable experimental performance. It is supplied as a lyophilized powder and has been verified via MALS and BLI for structural integrity and binding activity.
- Expressed in human 293 cells (HEK293)
- Contains amino acids Leu 20 to Pro 219 (Accession # AAI13679)
- Calculated molecular weight of 49.2 kDa, migrating as 60 to 70 kDa under reducing conditions due to glycosylation
- Endotoxin levels less than 0.01 EU per μg
- Purity ≥95% by SDS-PAGE and ≥90% by SEC-MALS
- Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose