Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human PD-L2 / B7-DC, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;PD2,PDL2,PD-L2,Butyrophilin B7-DC,CD273,PDCD1 ligand 2,PDCD1L2,PDCD1LG2,PDL-2,B7DC
Catalog No. 501659150
Encompass_Preferred
Recombinant Protein;Biotinylated Human PD-L2 / B7-DC, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human PD-L2, Fc Tag (PD2-H82F6) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession # Q9BQ51-1).;This protein carries a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 51 kDa. The protein migrates as 66-91 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Human PD-1, Fc Tag (Cat. No. PD1-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-L2, Fc Tag (Cat. No. PD2-H82F6) with a linear range of 0.5-16 ng/mL (QC tested).
Catalog No. 50-165-9150 Supplier ACROBiosystems Supplier No. PD2H82F625UG
Recombinant Protein;Biotinylated Human PD-L2 / B7-DC, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human PD-L2, Fc Tag (PD2-H82F6) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession # Q9BQ51-1).;This protein carries a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 51 kDa. The protein migrates as 66-91 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Human PD-1, Fc Tag (Cat. No. PD1-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-L2, Fc Tag (Cat. No. PD2-H82F6) with a linear range of 0.5-16 ng/mL (QC tested).