Learn More
ACROBiosystems Recombinant Protein;Mouse PD-L2 / B7-DC Protein;HEK293;100UG;PD2,PDL2,PD-L2,Butyrophilin B7-DC,CD273,PDCD1 ligand 2,PDCD1L2,PDCD1LG2,PDL-2,B7DC
Catalog No. 501658695
Encompass_Preferred
Recombinant Protein;Mouse PD-L2 / B7-DC Protein;100UG;Mouse PD-L2, His Tag (PD2-M52E3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 219 (Accession # NP_067371).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8695 Supplier ACROBiosystems Supplier No. PD2M52E3100UG
Recombinant Protein;Mouse PD-L2 / B7-DC Protein;100UG;Mouse PD-L2, His Tag (PD2-M52E3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 219 (Accession # NP_067371).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.