Learn More
BEZWADA BIOMEDICAL LLC Poly(D,L-lactide-co-glycolide), D,L-lactide:glycolide 50:50, viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904472
Encompass_Preferred
In order to decrease the absorption times of implantable devices incorporating L(-)-lactide, copolymers of L(-)-lactide and dl-lactide with glycolide have been prepared. The polymers are used in medical devices and drug delivery.
Catalog No. 50-190-4472 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 4255
In order to decrease the absorption times of implantable devices incorporating L(-)-lactide, copolymers of L(-)-lactide and dl-lactide with glycolide have been prepared. The polymers are used in medical devices and drug delivery.