Learn More
BEZWADA BIOMEDICAL LLC Poly(D,L-lactide-co-glycolide), D,L-lactide:glycolide 75:25, viscosity 0.55 to 0.75 dL/g, 5.0 GM
SDP
Catalog No. 501904476
Encompass_Preferred
In order to decrease the absorption times of implantable devices incorporating L(-)-lactide, copolymers of L(-)-lactide and dl-lactide with glycolide have been prepared. The polymers are used in medical devices and drug delivery.
Catalog No. 50-190-4476 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 4272
In order to decrease the absorption times of implantable devices incorporating L(-)-lactide, copolymers of L(-)-lactide and dl-lactide with glycolide have been prepared. The polymers are used in medical devices and drug delivery.