Learn More
Aviva Systems Biology Corp PDLIM1 Antibody - middle region (ARP37229_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37229_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PDLIM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse PDLIM1. Alternate names: Clim1, mClim1ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56340