Learn More
Aviva Systems Biology Corp PDLIM3 Antibody - N-terminal region (ARP55067_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55067_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDLIM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDLIM3. Alternate names: ALP, DKFZp686L0362ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61412