Learn More
Aviva Systems Biology Corp PDLIM5 Antibody - middle region (ARP34273_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34273_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PDLIM5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDLIM5. Alternate names: L9, ENH, LIM, ENH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55322