Learn More
Medchemexpress LLC HY-112629 10mg Medchemexpress, PDM2 CAS:688348-25-6 Purity:>98%
SDP
Catalog No. 501969123
Encompass_Preferred
Medchemexpress, HY-112629 10mg PDM2 CAS:688348-25-6 PDM2 is a selective, high-affinity aryl hydrocarbon receptor (AhR) antagonist with an Ki of 1.2±0.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9123 Supplier Medchemexpress LLC Supplier No. HY11262910MG
Medchemexpress, HY-112629 10mg PDM2 CAS:688348-25-6 PDM2 is a selective, high-affinity aryl hydrocarbon receptor (AhR) antagonist with an Ki of 1.2±0.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.