Learn More
AdipoGen PdPh4TBP
SDP
Catalog No. 502560021
Encompass_Preferred
Chemical. CAS 119654-64-7. Formula C60H36N4Pd. MW 919.392. Synthetic. Phosphorescent probe for measuring oxygen in very low concentrations. Luminescent marker for oxygen and pH in biomedical imaging. Used as sensitizer in the process of photon up-conversion based on triplet–triplet annihilation TTA. Used as agent for PDT, optical limiters and other types of nonlinear optical materials.
Catalog No. 50-256-0021 Supplier AdipoGen Supplier No. CDXT0083M025
Chemical. CAS 119654-64-7. Formula C60H36N4Pd. MW 919.392. Synthetic. Phosphorescent probe for measuring oxygen in very low concentrations. Luminescent marker for oxygen and pH in biomedical imaging. Used as sensitizer in the process of photon up-conversion based on triplet–triplet annihilation TTA. Used as agent for PDT, optical limiters and other types of nonlinear optical materials.