Learn More
Aviva Systems Biology Corp PDPK1 Antibody - middle region (ARP30313_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30313_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDPK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDPK1. Alternate names: MGC20087, MGC35290, PDK1, PRO0461, PkB-like, PkB-like 1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54153