Learn More
Aviva Systems Biology Corp PDPN Antibody - N-terminal region (ARP44528_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44528_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDPN. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PDPN. Alternate names: GP36, GP40, Gp38, HT1A-1, OTS8, PA2.26, T1A, T1A-2, AGGRUSApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-53427