Learn More
Worthington Biochemical Corporation PDS KIT INHIBITOR VIAL
SDP
Catalog No. 50592496
Encompass
Ovomucoid protease inhibitor and bovine serum albumin which is 0.22 micron filtered and lyophilized in autoclaved vials to contain 10 mg/ml each upon reconstitution with 32 ml of EBSS. 9035-81-8
Catalog No. 50-592-496 Supplier Worthington Biochemical Corporation Supplier No. LK003182
Ovomucoid protease inhibitor and bovine serum albumin which is 0.22 micron filtered and lyophilized in autoclaved vials to contain 10 mg/ml each upon reconstitution with 32 ml of EBSS. 9035-81-8