Learn More
BEZWADA BIOMEDICAL LLC Poly(P-dioxanone-co-glycolide), p-dioxanone:glycolide 90:10,viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904487
Encompass_Preferred
Copolymers of PDS with glycolide is prepared for Fast absorbion. Reduced absorption time from 3 months compared 9 months for PDS.BSR profile was reduced to 3 to 4 weeks in contrast to 6 to 8 weeks for PDS.No change in tensile properties, crystallinity, and thermal properties when compared to the PDS. Biomedical devices include surgical sutures, staples, meshes, and controlled drug delivery.
Catalog No. 50-190-4487 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 6291
Copolymers of PDS with glycolide is prepared for Fast absorbion. Reduced absorption time from 3 months compared 9 months for PDS.BSR profile was reduced to 3 to 4 weeks in contrast to 6 to 8 weeks for PDS.No change in tensile properties, crystallinity, and thermal properties when compared to the PDS. Biomedical devices include surgical sutures, staples, meshes, and controlled drug delivery.