Learn More
BEZWADA BIOMEDICAL LLC Poly(P-dioxanone-co-L-lactide), p-dioxanone:L-lactide 85:15, viscosity 1.5 to 2.5 dl/g, 5.0 GM
SDP
Catalog No. 501904490
Encompass_Preferred
Copolymers of PDS with L-lactide is soft and flexible and has exellent pliability. Biomedical devices include surgical sutures, staples, meshes, and controlled drug delivery.BSR profile longer than PDS.Biomedical devices include surgical sutures, staples, meshes, and controlled drug delivery.
Catalog No. 50-190-4490 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 6381
Copolymers of PDS with L-lactide is soft and flexible and has exellent pliability. Biomedical devices include surgical sutures, staples, meshes, and controlled drug delivery.BSR profile longer than PDS.Biomedical devices include surgical sutures, staples, meshes, and controlled drug delivery.