Learn More
Aviva Systems Biology Corp PDS5B Antibody - middle region (ARP58313_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58313_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDS5B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDS5B. Alternate names: APRIN, AS3, CG008, FLJ23236, KIAA0979, RP1-267P19.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62282