Learn More
Aviva Systems Biology Corp PDSS1 Antibody - middle region (ARP46196_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46196_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDSS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDSS1. Alternate names: COQ1, MGC70953, RP13-16H11.3, TPRT, TPT, hDPS1, DPS, SPS, TPT 1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59266