Learn More
Aviva Systems Biology Corp PDSS2 Antibody - C-terminal region (ARP48856_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48856_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDSS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PDSS2. Alternate names: C6orf210, DLP1, bA59I9.3, hDLP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59943