Learn More
Abcam PDTC (Ammonium pyrrolidinedithiocarbamate), NF-kappaB inhibitor antio x idant, 100MG
Catalog No. 502706643
Encompass_Preferred
MW 164.3 Da. Selective NF-κB inhibitor. Inhibits translation of nitric oxide synthase mRNA to prevent induction. Metal ion chelator and antioxidant. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6643 Supplier Abcam Supplier No. AB141406100MG
MW 164.3 Da. Selective NF-κB inhibitor. Inhibits translation of nitric oxide synthase mRNA to prevent induction. Metal ion chelator and antioxidant. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement