Learn More
Aviva Systems Biology Corp PDXK Antibody - N-terminal region (ARP53615_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53615_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDXK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PDXK. Alternate names: C21orf124, C21orf97, DKFZp566A071, FLJ31940, FLJ37311, MGC15873, MGC31754, MGC52346, PKH, PNK, PRED79ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-61070