Learn More
Aviva Systems Biology Corp PDXP Antibody - middle region (ARP57385_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57385_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDXP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDXP. Alternate names: CIN, FLJ32703, PLP, dJ37E16.5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-62127