Learn More
Aviva Systems Biology Corp Pdyn Antibody - middle region (ARP59573_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59573_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pdyn. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: DynApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-286-745