Learn More
Aviva Systems Biology Corp PDZK1 Antibody - N-terminal region (ARP41506_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41506_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDZK1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PDZK1. Alternate names: CAP70, CLAMP, NHERF3, PDZD1, NHERF-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-938