Learn More
Instech PE-10 tubing, .011x.024in (0.3x0.6mm), 30m (98ft) spool, non-sterile.
SDP
Catalog No. NC0695849
Encompass
The Instech BTPE-10 polyethylene tubing is PE-10 size (.011x.024in / 0.3x0.6mm) and supplied non-sterile on a 30m (98ft) spool. Its low‑density PE formulation provides excellent compound compatibility and low air permeability, making it suitable for general infusion and sampling applications. An economical alternative to Intramedic™ tubing, it is not recommended for intravascular use due to stiffness and should not be autoclaved.
Features
  • PE-10 size (.011x.024in / 0.3x0.6mm)
  • Low-density polyethylene (LDPE)
  • Excellent compound compatibility
  • Low air permeability
  • 30m non-sterile spool
  • Not for intravascular or human use
Catalog No. NC0695849 Supplier Instech Supplier No. BTPE10
The Instech BTPE-10 polyethylene tubing is PE-10 size (.011x.024in / 0.3x0.6mm) and supplied non-sterile on a 30m (98ft) spool. Its low‑density PE formulation provides excellent compound compatibility and low air permeability, making it suitable for general infusion and sampling applications. An economical alternative to Intramedic™ tubing, it is not recommended for intravascular use due to stiffness and should not be autoclaved.
Features
  • PE-10 size (.011x.024in / 0.3x0.6mm)
  • Low-density polyethylene (LDPE)
  • Excellent compound compatibility
  • Low air permeability
  • 30m non-sterile spool
  • Not for intravascular or human use