Learn More
Aat Bioquest PE [R-Phycoerythrin] *CAS 11016-17-4*
SDP
Supplier:  Aat Bioquest 2557
Encompass_Preferred
R-Phycoerythrin (PE) is isolated from red algae. Its primary absorption peak is at 565 nm with secondary peaks at 496 and 545 nm. The relative prominence of the secondary peaks varies significantly among R-PEs from different species.
Catalog No. 50-195-3186