Learn More
Perkin Elmer US LLC Elite-23 Capillary Column - 30 m x 0.25 mm I.D. x 0.25 µm
Catalog No. 502016592
Encompass_Preferred
For the analysis of cis/trans isomers in FAMEs and dioxins at temperatures ranging from 0°C to 275°C. Equivalent to USP G5.
Catalog No. 50-201-6592 Supplier Perkin Elmer US LLC Supplier No. N9316506
For the analysis of cis/trans isomers in FAMEs and dioxins at temperatures ranging from 0°C to 275°C. Equivalent to USP G5.