Learn More
Perkin Elmer US LLC PE 2400 Series Maintenance Kit
Catalog No. 502104305
Encompass_Preferred
Included in this kit are: Washer O-Rings, Qty. 3 Skinner O-Ring, Qty. 1 Black Viton O-Rings, 0.421 in ID, Qty. 3 Black Viton O-Rings, 0.859 in, Qty. 3 Viton O-Rings, 0.614 in ID, Qty. 2 Viton O-Ring, 3.487 in ID, Qty. 1 Red Silicone O-Rings, 0.859 in ID, Qty. 2 Solenoid Gas Line Filter, Qty. 1 Stubby Spring Plunger, Qty. 1 Solenoid Open Valve Accessory, Qty. 1 Glass Tube Washers, 2 mm, Qty. 2 Normally Closed Valve Plungers, Qty. 5
Catalog No. 50-210-4305 Supplier Perkin Elmer US LLC Supplier No. N2410152
Included in this kit are: Washer O-Rings, Qty. 3 Skinner O-Ring, Qty. 1 Black Viton O-Rings, 0.421 in ID, Qty. 3 Black Viton O-Rings, 0.859 in, Qty. 3 Viton O-Rings, 0.614 in ID, Qty. 2 Viton O-Ring, 3.487 in ID, Qty. 1 Red Silicone O-Rings, 0.859 in ID, Qty. 2 Solenoid Gas Line Filter, Qty. 1 Stubby Spring Plunger, Qty. 1 Solenoid Open Valve Accessory, Qty. 1 Glass Tube Washers, 2 mm, Qty. 2 Normally Closed Valve Plungers, Qty. 5