Learn More
Biolegend PE anti-CD247 (TCRζ, CD3ζ)
Supplier:  Biolegend 644106
Encompass_Preferred
PE anti-CD247 (TCRζ, CD3ζ) [6B10.2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 ug
Catalog No. NC0944700