Learn More
Biolegend PE anti-human/mouse CD3ε (activation epitope)
Supplier:  Biolegend 362703
Encompass_Preferred
PE anti-human/mouse CD3ε (activation epitope) [APA1/1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 25 tests
Catalog No. 50-207-8085