Learn More
Biolegend PE anti-Cyclin B1
Supplier:  Biolegend 647904
Encompass_Preferred
PE anti-Cyclin B1 [V152]; Isotype: Mouse IgG1; Reactivity: Human, Cross-Reactivity: Mouse; Apps: ICFC; Size: 100 tests
Catalog No. 50-208-6956