Learn More
Biolegend PE anti-GATA3
Supplier:  Biolegend 653804
Encompass_Preferred
PE anti-GATA3 [16E10A23]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests
Catalog No. 50-604-767