Learn More
Biolegend PE anti-H2A.X Phospho (Ser139)
Supplier:  Biolegend 613411
Encompass_Preferred
PE anti-H2A.X Phospho (Ser139) [2F3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-6718