Learn More
Biolegend PE anti-HDAC1 Phospho (Ser406)
Supplier:  Biolegend 611004
Encompass_Preferred
PE anti-HDAC1 Phospho (Ser406) [BT-15]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Zebrafish; Apps: ICFC; Size: 100 tests
Catalog No. 50-208-6701