Learn More
Biolegend PE anti-human C5L2
Supplier:  Biolegend 342404
Encompass_Preferred
PE anti-human C5L2 [1D9-M12]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-168-127