Learn More
Biolegend PE anti-human CD105
Supplier:  Biolegend 323206
Encompass_Preferred
PE anti-human CD105 [43A3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-174