Learn More
Biolegend PE anti-human CD106
Supplier:  Biolegend 305806
Encompass_Preferred
PE anti-human CD106 [STA]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-166-302