Learn More
Biolegend PE anti-human CD123
Supplier:  Biolegend 306005
Encompass_Preferred
PE anti-human CD123 [6H6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-311