Learn More
Biolegend PE anti-human CD130 (gp130)
Supplier:  Biolegend 362004
Encompass_Preferred
PE anti-human CD130 (gp130) [2E1B02]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-210