Learn More
Biolegend PE anti-human CD14
Supplier:  Biolegend 325606
Encompass_Preferred
PE anti-human CD14 [HCD14]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-292